Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286431.1 | NP_001273360.1 | 34 - 35 | 9951 | 68 |
NM_001286432.1 | NP_001273361.1 | 33 - 34 | 9844 | 68 | |
NM_002912.4 | NP_002903.3 | 32 - 33 | 9716 | 68 | |
XM_011536028.1 | XP_011534330.1 | 32 - 33 | 9888 | 68 | |
XM_011536029.2 | XP_011534331.1 | 32 - 33 | 9885 | 68 | |
XM_011536030.2 | XP_011534332.1 | 31 - 32 | 9810 | 68 | |
XM_011536031.2 | XP_011534333.1 | 33 - 34 | 10016 | 68 | |
XM_011536032.2 | XP_011534334.1 | 33 - 34 | 14606 | 68 | |
XM_017011152.1 | XP_016866641.1 | 33 - 34 | 10013 | 68 | |
XM_017011153.1 | XP_016866642.1 | 33 - 34 | 9556 | 68 | |
XM_017011154.1 | XP_016866643.1 | 33 - 34 | 9441 | 68 | |
GenBank mRNA | AB209256.1 | - | 20 - 21 | 3770 | 68 |
AF004713.1 | - | 6 - 7 | 937 | 68 | |
AF035537.1 | - | 30 - 31 | 9053 | 68 | |
AF058701.1 | - | 32 - 33 | 9343 | 68 | |
AF071798.1 | - | 31 - 32 | 9581 | 68 | |
AF078695.1 | - | 33 - 34 | 9846 | 68 | |
AF157476.1 | - | 31 - 32 | 9289 | 68 | |
AF179428.1 | - | 32 - 33 | 9417 | 68 | |
AF179429.1 | - | 33 - 34 | 9524 | 68 | |
AL096744.1 | - | 6 - 7 | 1395 | 68 |