Assay Design



Gene Transcripts

Gene Symbol
RPS6KA4
Entrez Gene ID
8986
Gene Name
ribosomal protein S6 kinase A4
Gene Aliases
MSK2, RSK-B, S6K-alpha-4
Chromosome Location
Chr.11: 64359153 - 64372215 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006944.1 NP_001006945.1 10 - 11 1285 141
GenBank mRNA AF074715.1 - 9 - 10 1154 141

View Full Product Details