Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271641.1 | NP_001258570.1 | 7 - 8 | 1137 | 98 |
NM_014341.2 | NP_055156.1 | 7 - 8 | 1137 | 98 | |
NR_130739.1 | 7 - 8 | 1230 | 98 | ||
XM_005248978.1 | XP_005249035.1 | 7 - 8 | 1137 | 98 | |
XM_005248979.1 | XP_005249036.1 | 7 - 8 | 1137 | 98 | |
XM_011514450.1 | XP_011512752.1 | 6 - 7 | 645 | 98 | |
XM_017010642.1 | XP_016866131.1 | 6 - 7 | 645 | 98 | |
XM_017010643.1 | XP_016866132.1 | 6 - 7 | 640 | 98 | |
GenBank mRNA | AF151822.1 | - | 7 - 8 | 609 | 98 |
AF176006.3 | - | 7 - 8 | 759 | 98 | |
AF189289.1 | - | 7 - 8 | 718 | 98 | |
AF192559.3 | - | 7 - 8 | 759 | 98 | |
AK022783.1 | - | 7 - 8 | 757 | 98 | |
AK130842.1 | - | 7 - 8 | 841 | 98 | |
AK225628.1 | - | 7 - 8 | 740 | 98 | |
AK303318.1 | - | 6 - 7 | 558 | 98 | |
AK315067.1 | - | 7 - 8 | 1138 | 98 | |
AY358519.1 | - | 4 - 5 | 218 | 98 | |
AY947344.1 | - | 7 - 8 | 728 | 98 | |
BC000702.2 | - | 7 - 8 | 689 | 98 | |
BC006507.2 | - | 7 - 8 | 489 | 98 | |
BC009962.2 | - | 7 - 8 | 738 | 98 | |
BC040659.1 | - | 7 - 8 | 705 | 98 | |
BC110914.1 | - | 7 - 8 | 780 | 98 | |
BC125056.1 | - | 7 - 8 | 754 | 98 | |
BC153873.1 | - | 7 - 8 | 759 | 98 |