Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145038.4 | NP_659475.2 | 8 - 9 | 1106 | 153 |
XM_005264637.3 | XP_005264694.1 | 7 - 8 | 914 | 153 | |
GenBank mRNA | AK057222.1 | - | 8 - 9 | 1102 | 153 |
AL833892.1 | - | 7 - 8 | 840 | 153 | |
BC009488.2 | - | 7 - 8 | 829 | 153 |