Assay Design



Gene Transcripts

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Chromosome Location
Chr.17: 46029916 - 46225374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193466.1 NP_001180395.1 10 - 11 3011 66
NM_015443.3 NP_056258.1 10 - 11 2713 66
XM_006721823.1 XP_006721886.1 11 - 12 2797 66
XM_006721824.3 XP_006721887.1 11 - 12 2981 66
XM_011524629.2 XP_011522931.1 10 - 11 2879 66
XM_011524630.2 XP_011522932.1 10 - 11 2792 66
XM_011524632.2 XP_011522934.1 10 - 11 1407 66
GenBank mRNA AB033093.1 - 9 - 10 2634 66
AK094946.1 - 2 - 3 512 66
AL137317.1 - 6 - 7 955 66
BC035892.1 - 5 - 6 818 66
BC098376.1 - 10 - 11 2713 66
BX538006.1 - 10 - 11 3014 66
BX648760.1 - 3 - 4 6771 66

View Full Product Details