Assay Design



Gene Transcripts

Gene Symbol
LETMD1
Entrez Gene ID
25875
Gene Name
LETM1 domain containing 1
Gene Aliases
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location
Chr.12: 51048230 - 51068984 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243689.1 NP_001230618.1 2 - 3 331 155
NM_001300765.1 NP_001287694.1 2 - 3 331 155
NM_015416.4 NP_056231.3 2 - 3 331 155
NR_045017.1 2 - 3 206 155
XM_006719339.1 XP_006719402.1 2 - 3 303 155
XM_011538161.2 XP_011536463.1 2 - 3 338 155
XM_011538162.2 XP_011536464.1 2 - 3 339 155
XM_011538163.2 XP_011536465.1 2 - 3 340 155
XM_011538165.2 XP_011536467.1 2 - 3 338 155
XM_017019151.1 XP_016874640.1 2 - 3 340 155
XM_017019152.1 XP_016874641.1 2 - 3 306 155
XM_017019155.1 XP_016874644.1 2 - 3 178 155
XM_017019159.1 XP_016874648.1 2 - 3 341 155
XM_017019160.1 XP_016874649.1 2 - 3 303 155
XM_017019165.1 XP_016874654.1 2 - 3 303 155
XM_017019167.1 XP_016874656.1 2 - 3 303 155
XM_017019168.1 XP_016874657.1 2 - 3 303 155
GenBank mRNA AF195651.2 - 2 - 3 303 155
AF315598.1 - 2 - 3 167 155
AK000639.1 - 2 - 3 318 155
AK057866.1 - 2 - 3 276 155
AK127540.1 - 2 - 3 292 155
AK225836.1 - 2 - 3 289 155
AK315119.1 - 2 - 3 155
BC064943.1 - 2 - 3 277 155
BX376670.2 - 2 - 3 272 155
DC357852.1 - 2 - 3 331 155

View Full Product Details