Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015416.4 | NP_056231.3 | 3 - 4 | 451 | 89 |
NR_045017.1 | 3 - 4 | 326 | 89 | ||
XM_011538161.2 | XP_011536463.1 | 3 - 4 | 458 | 89 | |
XM_011538162.2 | XP_011536464.1 | 3 - 4 | 459 | 89 | |
XM_011538163.2 | XP_011536465.1 | 3 - 4 | 460 | 89 | |
XM_017019151.1 | XP_016874640.1 | 3 - 4 | 460 | 89 | |
XM_017019152.1 | XP_016874641.1 | 3 - 4 | 426 | 89 | |
XM_017019155.1 | XP_016874644.1 | 3 - 4 | 298 | 89 | |
GenBank mRNA | AF195651.2 | - | 3 - 4 | 423 | 89 |
AF315598.1 | - | 3 - 4 | 287 | 89 | |
AK000639.1 | - | 3 - 4 | 438 | 89 | |
AK057866.1 | - | 3 - 4 | 396 | 89 | |
AK225836.1 | - | 3 - 4 | 409 | 89 | |
AK301955.1 | - | 3 - 4 | 326 | 89 | |
AK315119.1 | - | 3 - 4 | 418 | 89 | |
BC064943.1 | - | 3 - 4 | 397 | 89 | |
DC357852.1 | - | 3 - 4 | 451 | 89 |