Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164741.1 | NP_001158213.1 | 2 - 3 | 331 | 167 |
NM_001666.4 | NP_001657.3 | 2 - 3 | 331 | 167 | |
GenBank mRNA | AK294562.1 | - | 2 - 3 | 256 | 167 |
AY183123.1 | - | 2 - 3 | 315 | 167 | |
BC052303.1 | - | 2 - 3 | 331 | 167 | |
D50921.1 | - | 2 - 3 | 262 | 167 | |
X78817.1 | - | 2 - 3 | 315 | 167 |