Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005256437.2 | XP_005256494.1 | 17 - 18 | 3401 | 65 |
XM_005256438.2 | XP_005256495.1 | 17 - 18 | 3385 | 65 | |
XM_017024130.1 | XP_016879619.1 | 18 - 19 | 3257 | 65 | |
XM_017024131.1 | XP_016879620.1 | 18 - 19 | 2745 | 65 | |
XM_017024136.1 | XP_016879625.1 | 17 - 18 | 3374 | 65 | |
XM_017024139.1 | XP_016879628.1 | 17 - 18 | 3471 | 65 | |
GenBank mRNA | BC014055.2 | - | 16 - 17 | 2212 | 65 |