Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130447.1 | NP_001123919.1 | 2 - 3 | 229 | 124 |
NM_018097.2 | NP_060567.1 | 3 - 4 | 322 | 124 | |
GenBank mRNA | AK001322.1 | - | 3 - 4 | 280 | 124 |
BC010903.1 | - | 3 - 4 | 301 | 124 | |
DA923763.1 | - | 2 - 3 | 186 | 124 | |
DB450370.1 | - | 3 - 4 | 322 | 124 |