Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 10 - 11 | 1251 | 82 |
NM_013241.2 | NP_037373.2 | 10 - 11 | 1251 | 82 | |
XM_011523043.2 | XP_011521345.1 | 10 - 11 | 1467 | 82 | |
XM_011523044.1 | XP_011521346.1 | 10 - 11 | 1196 | 82 | |
XM_011523045.2 | XP_011521347.1 | 10 - 11 | 961 | 82 | |
GenBank mRNA | AB041046.1 | - | 10 - 11 | 1221 | 82 |
AB209584.1 | - | 8 - 9 | 1720 | 82 | |
AF113615.1 | - | 10 - 11 | 1157 | 82 | |
BC033084.1 | - | 10 - 11 | 1196 | 82 | |
KJ658417.1 | - | 10 - 11 | 1139 | 82 |