Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013241.2 | NP_037373.2 | 11 - 12 | 1429 | 68 |
GenBank mRNA | AB041046.1 | - | 11 - 12 | 1399 | 68 |
AB209584.1 | - | 9 - 10 | 1898 | 68 | |
AF113615.1 | - | 11 - 12 | 1335 | 68 | |
AK304232.1 | - | 11 - 12 | 1364 | 68 | |
AY192154.1 | - | 11 - 12 | 1317 | 68 | |
BC033084.1 | - | 11 - 12 | 1374 | 68 |