Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 17 - 18 | 2575 | 72 |
NM_013241.2 | NP_037373.2 | 15 - 16 | 2497 | 72 | |
XM_006721180.1 | XP_006721243.1 | 6 - 7 | 1150 | 72 | |
XM_011523043.2 | XP_011521345.1 | 17 - 18 | 2791 | 72 | |
XM_011523044.1 | XP_011521346.1 | 17 - 18 | 2520 | 72 | |
XM_011523045.2 | XP_011521347.1 | 17 - 18 | 2285 | 72 | |
GenBank mRNA | AB041046.1 | - | 15 - 16 | 2467 | 72 |
AB209584.1 | - | 13 - 14 | 2966 | 72 | |
AF113615.1 | - | 15 - 16 | 2403 | 72 | |
AK025992.1 | - | 1 - 2 | 187 | 72 | |
AK301157.1 | - | 5 - 6 | 1208 | 72 | |
AY192154.1 | - | 15 - 16 | 2385 | 72 | |
BC033084.1 | - | 15 - 16 | 2442 | 72 | |
KJ658417.1 | - | 17 - 18 | 2463 | 72 |