Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319209.1 | NP_001306138.1 | 2 - 3 | 273 | 79 |
NM_002722.4 | NP_002713.1 | 2 - 3 | 232 | 79 | |
XM_011524978.2 | XP_011523280.1 | 2 - 3 | 386 | 79 | |
GenBank mRNA | BC032225.2 | - | - | 185 | 79 |
BC040033.2 | - | 1 - 2 | 185 | 79 | |
BP384893.1 | - | 2 - 3 | 273 | 79 | |
BP388331.1 | - | 1 - 2 | 109 | 79 | |
BP394217.1 | - | 2 - 3 | 275 | 79 | |
M15788.1 | - | 1 - 2 | 193 | 79 | |
X00491.1 | - | 1 - 2 | 199 | 79 |