Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000321.2 | NP_000312.2 | 17 - 18 | 1862 | 126 |
XM_011535171.2 | XP_011533473.1 | 16 - 17 | 1451 | 126 | |
GenBank mRNA | AB208788.1 | - | 17 - 18 | 1793 | 126 |
AK291258.1 | - | 17 - 18 | 1883 | 126 | |
AK299179.1 | - | 18 - 19 | 1773 | 126 | |
AK300284.1 | - | 18 - 19 | 1799 | 126 | |
AK307730.1 | - | 17 - 18 | 1849 | 126 | |
BC039060.1 | - | 17 - 18 | 1848 | 126 | |
BC040540.1 | - | 17 - 18 | 1858 | 126 | |
L41870.1 | - | 17 - 18 | 1833 | 126 | |
M15400.1 | - | 17 - 18 | 1834 | 126 | |
M28419.1 | - | 17 - 18 | 1834 | 126 | |
M33647.1 | - | 17 - 18 | 1699 | 126 |