Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322194.1 | NP_001309123.1 | 12 - 13 | 2240 | 122 |
NM_001322195.1 | NP_001309124.1 | 11 - 12 | 1758 | 122 | |
NM_001322196.1 | NP_001309125.1 | 11 - 12 | 2071 | 122 | |
NM_001322198.1 | NP_001309127.1 | 12 - 13 | 2059 | 122 | |
NM_001322199.1 | NP_001309128.1 | 12 - 13 | 2001 | 122 | |
NM_001322204.1 | NP_001309133.1 | 9 - 10 | 1510 | 122 | |
NM_004972.3 | NP_004963.1 | 12 - 13 | 2139 | 122 | |
GenBank mRNA | AF005216.1 | - | 12 - 13 | 2139 | 122 |
AF058925.1 | - | 12 - 13 | 2122 | 122 | |
AK292525.1 | - | 11 - 12 | 1758 | 122 | |
AK302618.1 | - | 9 - 10 | 1569 | 122 | |
AY973034.1 | - | 11 - 12 | 1716 | 122 | |
BC039695.1 | - | 11 - 12 | 1743 | 122 |