Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004869.3 | NP_004860.2 | 7 - 8 | 1050 | 79 |
XM_006722582.2 | XP_006722645.1 | 6 - 7 | 926 | 79 | |
XM_011526268.2 | XP_011524570.1 | 7 - 8 | 1050 | 79 | |
GenBank mRNA | AF038960.1 | - | 7 - 8 | 979 | 79 |
AF195514.1 | - | 7 - 8 | 991 | 79 | |
AK290932.1 | - | 7 - 8 | 994 | 79 | |
AK291253.1 | - | 7 - 8 | 948 | 79 | |
AK308892.1 | - | 7 - 8 | 1014 | 79 | |
AK309322.1 | - | 6 - 7 | 926 | 79 | |
AK311041.1 | - | 6 - 7 | 926 | 79 | |
AY232629.1 | - | 7 - 8 | 800 | 79 | |
BC039574.1 | - | 7 - 8 | 957 | 79 |