Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080480.2 | NP_001073949.1 | 13 - 13 | 3003 | 138 |
NR_073465.1 | 10 - 10 | 2723 | 138 | ||
XM_006714999.2 | XP_006715062.1 | 13 - 13 | 2740 | 138 | |
GenBank mRNA | AK093994.1 | - | 7 - 7 | 2198 | 138 |