Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 17 - 18 | 2647 | 92 |
XM_005268497.1 | XP_005268554.1 | 17 - 18 | 2611 | 92 | |
XM_005268498.1 | XP_005268555.1 | 17 - 18 | 2611 | 92 | |
XM_005268499.1 | XP_005268556.1 | 17 - 18 | 2585 | 92 | |
XM_005268500.3 | XP_005268557.1 | 17 - 18 | 2611 | 92 | |
XM_006714792.1 | XP_006714855.1 | 17 - 18 | 2611 | 92 | |
XM_011537677.2 | XP_011535979.1 | 15 - 16 | 2066 | 92 | |
GenBank mRNA | AJ310567.1 | - | 16 - 17 | 2597 | 92 |
AK001579.1 | - | 1 - 2 | 318 | 92 | |
AK091385.1 | - | 17 - 18 | 2526 | 92 | |
AK290052.1 | - | 17 - 18 | 92 | ||
AK295768.1 | - | 11 - 12 | 1624 | 92 |