Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 26 - 27 | 3733 | 57 |
XM_005268497.1 | XP_005268554.1 | 26 - 27 | 3697 | 57 | |
XM_005268498.1 | XP_005268555.1 | 26 - 27 | 3637 | 57 | |
XM_005268499.1 | XP_005268556.1 | 26 - 27 | 3671 | 57 | |
XM_006714792.1 | XP_006714855.1 | 25 - 26 | 3484 | 57 | |
XM_011537677.2 | XP_011535979.1 | 24 - 25 | 3152 | 57 | |
GenBank mRNA | AJ310567.1 | - | 25 - 26 | 3683 | 57 |
AK001579.1 | - | 9 - 10 | 1657 | 57 | |
AK091385.1 | - | 25 - 26 | 3399 | 57 | |
AK290052.1 | - | 26 - 27 | 3726 | 57 | |
AK295768.1 | - | 20 - 21 | 2710 | 57 |