Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 29 - 30 | 3986 | 104 |
XM_005268497.1 | XP_005268554.1 | 29 - 30 | 3950 | 104 | |
XM_005268498.1 | XP_005268555.1 | 29 - 30 | 3890 | 104 | |
XM_005268499.1 | XP_005268556.1 | 29 - 30 | 3924 | 104 | |
XM_006714792.1 | XP_006714855.1 | 28 - 29 | 3737 | 104 | |
XM_011537677.2 | XP_011535979.1 | 27 - 28 | 3405 | 104 | |
GenBank mRNA | AJ310567.1 | - | 28 - 29 | 3936 | 104 |
AK001579.1 | - | 12 - 13 | 1910 | 104 | |
AK091385.1 | - | 28 - 29 | 3652 | 104 | |
AK290052.1 | - | 29 - 30 | 3979 | 104 | |
AK295768.1 | - | 23 - 24 | 2963 | 104 | |
BC009968.2 | - | 3 - 4 | 237 | 104 |