Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 30 - 31 | 4055 | 69 |
XM_005268497.1 | XP_005268554.1 | 30 - 31 | 4019 | 69 | |
XM_005268498.1 | XP_005268555.1 | 30 - 31 | 3959 | 69 | |
XM_005268499.1 | XP_005268556.1 | 30 - 31 | 3993 | 69 | |
XM_006714792.1 | XP_006714855.1 | 29 - 30 | 3806 | 69 | |
XM_011537677.2 | XP_011535979.1 | 28 - 29 | 3474 | 69 | |
GenBank mRNA | AJ310567.1 | - | 29 - 30 | 4005 | 69 |
AK001579.1 | - | 13 - 14 | 1979 | 69 | |
AK091385.1 | - | 29 - 30 | 3721 | 69 | |
AK290052.1 | - | 30 - 31 | 4048 | 69 | |
AK295768.1 | - | 24 - 25 | 3032 | 69 | |
BC009968.2 | - | 4 - 5 | 306 | 69 |