Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004586.2 | NP_004577.1 | 13 - 14 | 1096 | 149 |
XM_005274573.2 | XP_005274630.1 | 13 - 14 | 1100 | 149 | |
XM_005274577.2 | XP_005274634.1 | 12 - 13 | 1036 | 149 | |
XM_006724507.3 | XP_006724570.1 | 15 - 16 | 1202 | 149 | |
XM_011545555.1 | XP_011543857.1 | 13 - 14 | 1118 | 149 | |
XM_011545556.1 | XP_011543858.1 | 13 - 14 | 1118 | 149 | |
XM_011545557.2 | XP_011543859.1 | 15 - 16 | 1236 | 149 | |
XM_011545558.2 | XP_011543860.1 | 15 - 16 | 1300 | 149 | |
XM_011545559.1 | XP_011543861.1 | 15 - 16 | 1297 | 149 | |
XM_011545560.1 | XP_011543862.1 | 14 - 15 | 1152 | 149 | |
XM_011545561.1 | XP_011543863.1 | 13 - 14 | 1078 | 149 | |
XM_011545562.1 | XP_011543864.1 | 12 - 13 | 1054 | 149 | |
XM_011545563.2 | XP_011543865.1 | 15 - 16 | 1216 | 149 | |
XM_017029713.1 | XP_016885202.1 | 15 - 16 | 1204 | 149 | |
XM_017029714.1 | XP_016885203.1 | 15 - 16 | 1280 | 149 | |
XM_017029715.1 | XP_016885204.1 | 15 - 16 | 1279 | 149 | |
XM_017029716.1 | XP_016885205.1 | 15 - 16 | 1354 | 149 | |
XM_017029717.1 | XP_016885206.1 | 13 - 14 | 1060 | 149 | |
XM_017029718.1 | XP_016885207.1 | 12 - 13 | 1036 | 149 | |
XM_017029719.1 | XP_016885208.1 | 13 - 14 | 1060 | 149 | |
GenBank mRNA | AB102662.1 | - | 13 - 14 | 1055 | 149 |
AB208933.1 | - | 13 - 14 | 1298 | 149 | |
AK294373.1 | - | 15 - 16 | 1191 | 149 | |
AK296775.1 | - | 12 - 13 | 1032 | 149 | |
AK313932.1 | - | 13 - 14 | 1100 | 149 | |
AK316482.1 | - | 14 - 15 | 1495 | 149 | |
BC096301.1 | - | 13 - 14 | 1096 | 149 | |
BC096302.3 | - | 13 - 14 | 1096 | 149 | |
BC096303.1 | - | 13 - 14 | 1096 | 149 | |
L07599.1 | - | 13 - 14 | 1093 | 149 | |
U08316.1 | - | 13 - 14 | 1096 | 149 |