Assay Design



Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291339.1 NP_001278268.1 18 - 19 2722 107
NM_024790.6 NP_079066.5 21 - 22 2722 107
XM_005251305.3 XP_005251362.2 24 - 25 3206 107
XM_006716474.2 XP_006716537.2 23 - 24 3053 107
XM_006716477.2 XP_006716540.2 19 - 20 2675 107
XM_011517598.1 XP_011515900.1 25 - 26 3248 107
XM_011517599.1 XP_011515901.1 24 - 25 3224 107
XM_011517600.1 XP_011515902.1 23 - 24 3182 107
XM_011517601.1 XP_011515903.1 23 - 24 3143 107
XM_011517602.1 XP_011515904.1 22 - 23 3101 107
XM_011517603.1 XP_011515905.1 24 - 25 2931 107
XM_011517607.1 XP_011515909.1 23 - 24 2811 107
XM_011517608.1 XP_011515910.1 22 - 23 2808 107
XM_011517609.1 XP_011515911.1 21 - 22 2930 107
XM_011517611.2 XP_011515913.1 9 - 10 1307 107
XM_017013847.1 XP_016869336.1 23 - 24 3107 107
XM_017013848.1 XP_016869337.1 22 - 23 3083 107
XM_017013849.1 XP_016869338.1 22 - 23 3044 107
XM_017013850.1 XP_016869339.1 21 - 22 3002 107
XM_017013851.1 XP_016869340.1 21 - 22 2766 107
XM_017013852.1 XP_016869341.1 20 - 21 2849 107
XM_017013853.1 XP_016869342.1 20 - 21 2613 107
XM_017013854.1 XP_016869343.1 18 - 19 2651 107
XM_017013855.1 XP_016869344.1 16 - 17 2417 107
XM_017013856.1 XP_016869345.1 16 - 17 2235 107
XM_017013857.1 XP_016869346.1 17 - 18 2285 107
XM_017013858.1 XP_016869347.1 15 - 16 2033 107
XM_017013859.1 XP_016869348.1 11 - 12 1435 107
GenBank mRNA AK026143.1 - - 169 107
BC022867.1 - 3 - 4 221 107
BX649147.1 - 21 - 22 2723 108

View Full Product Details