Assay Design



Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024790.6 NP_079066.5 3 - 4 334 97
XM_005251305.3 XP_005251362.2 4 - 5 713 97
XM_006716474.2 XP_006716537.2 4 - 5 713 97
XM_006716477.2 XP_006716540.2 4 - 5 713 97
XM_011517598.1 XP_011515900.1 4 - 5 713 97
XM_011517599.1 XP_011515901.1 4 - 5 713 97
XM_011517600.1 XP_011515902.1 4 - 5 713 97
XM_011517601.1 XP_011515903.1 4 - 5 713 97
XM_011517602.1 XP_011515904.1 4 - 5 713 97
XM_011517603.1 XP_011515905.1 3 - 4 396 97
XM_011517607.1 XP_011515909.1 3 - 4 300 97
XM_011517608.1 XP_011515910.1 3 - 4 378 97
XM_017013847.1 XP_016869336.1 4 - 5 713 97
XM_017013848.1 XP_016869337.1 4 - 5 713 97
XM_017013849.1 XP_016869338.1 4 - 5 713 97
XM_017013850.1 XP_016869339.1 4 - 5 713 97
XM_017013851.1 XP_016869340.1 3 - 4 378 97
XM_017013852.1 XP_016869341.1 4 - 5 713 97
XM_017013853.1 XP_016869342.1 3 - 4 378 97
XM_017013854.1 XP_016869343.1 4 - 5 713 97
XM_017013855.1 XP_016869344.1 4 - 5 713 97
XM_017013856.1 XP_016869345.1 3 - 4 378 97
GenBank mRNA AK096205.1 - 3 - 4 307 97
AK127108.1 - 3 - 4 274 97
AM156947.1 - 3 - 4 335 97
BX649147.1 - 3 - 4 335 97
DA245734.1 - 3 - 4 275 97

View Full Product Details