Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291339.1 | NP_001278268.1 | 5 - 6 | 1240 | 144 |
NM_024790.6 | NP_079066.5 | 7 - 8 | 1087 | 144 | |
XM_005251305.3 | XP_005251362.2 | 10 - 11 | 1571 | 144 | |
XM_006716474.2 | XP_006716537.2 | 10 - 11 | 1571 | 144 | |
XM_011517598.1 | XP_011515900.1 | 10 - 11 | 1571 | 144 | |
XM_011517599.1 | XP_011515901.1 | 9 - 10 | 1547 | 144 | |
XM_011517600.1 | XP_011515902.1 | 9 - 10 | 1547 | 144 | |
XM_011517601.1 | XP_011515903.1 | 8 - 9 | 1466 | 144 | |
XM_011517602.1 | XP_011515904.1 | 8 - 9 | 1466 | 144 | |
XM_011517603.1 | XP_011515905.1 | 9 - 10 | 1254 | 144 | |
XM_011517607.1 | XP_011515909.1 | 8 - 9 | 1134 | 144 | |
XM_011517608.1 | XP_011515910.1 | 7 - 8 | 1131 | 144 | |
XM_011517609.1 | XP_011515911.1 | 6 - 7 | 1253 | 144 | |
XM_017013851.1 | XP_016869340.1 | 7 - 8 | 1131 | 144 | |
XM_017013853.1 | XP_016869342.1 | 7 - 8 | 1131 | 144 | |
XM_017013857.1 | XP_016869346.1 | 3 - 4 | 640 | 144 | |
GenBank mRNA | AJ583433.1 | - | 5 - 6 | 1240 | 144 |
AK096205.1 | - | 8 - 9 | 1141 | 144 | |
AK127108.1 | - | 9 - 10 | 1132 | 144 | |
AM156947.1 | - | 7 - 8 | 1088 | 144 | |
BX649147.1 | - | 7 - 8 | 1088 | 144 |