Assay Design



Gene Transcripts

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
Chromosome Location
Chr.8: 67062417 - 67196614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024790.6 NP_079066.5 15 - 16 1989 88
XM_005251305.3 XP_005251362.2 18 - 19 2473 88
XM_006716477.2 XP_006716540.2 13 - 14 1942 88
XM_011517598.1 XP_011515900.1 19 - 20 2515 88
XM_011517599.1 XP_011515901.1 18 - 19 2491 88
XM_011517600.1 XP_011515902.1 17 - 18 2449 88
XM_011517601.1 XP_011515903.1 17 - 18 2410 88
XM_011517602.1 XP_011515904.1 16 - 17 2368 88
XM_011517603.1 XP_011515905.1 18 - 19 2198 88
XM_011517607.1 XP_011515909.1 17 - 18 2078 88
XM_011517608.1 XP_011515910.1 16 - 17 2075 88
XM_011517609.1 XP_011515911.1 15 - 16 2197 88
XM_011517611.2 XP_011515913.1 3 - 4 574 88
XM_017013847.1 XP_016869336.1 17 - 18 2374 88
XM_017013848.1 XP_016869337.1 16 - 17 2350 88
XM_017013849.1 XP_016869338.1 16 - 17 2311 88
XM_017013850.1 XP_016869339.1 15 - 16 2269 88
XM_017013851.1 XP_016869340.1 15 - 16 2033 88
XM_017013854.1 XP_016869343.1 12 - 13 1918 88
XM_017013856.1 XP_016869345.1 10 - 11 1502 88
XM_017013857.1 XP_016869346.1 11 - 12 1552 88
XM_017013859.1 XP_016869348.1 5 - 6 702 88
GenBank mRNA AK096205.1 - 16 - 17 2043 88
AK127108.1 - 18 - 19 3518 88
AM156947.1 - 15 - 16 1990 88
BX649147.1 - 15 - 16 1990 88

View Full Product Details