Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005392.3 | NP_005383.3 | 10 - 11 | 1385 | 55 |
XM_005252051.2 | XP_005252108.1 | 10 - 11 | 1379 | 55 | |
XM_006717143.2 | XP_006717206.1 | 10 - 11 | 1274 | 55 | |
GenBank mRNA | AB014562.2 | - | 9 - 10 | 1278 | 55 |
AF043725.1 | - | 10 - 11 | 1592 | 55 | |
AL834263.1 | - | 10 - 11 | 1236 | 55 | |
BC152437.1 | - | 9 - 10 | 1278 | 55 |