Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031685.2 | NP_001026855.2 | 14 - 15 | 3149 | 84 |
NM_005426.2 | NP_005417.1 | 15 - 16 | 3281 | 84 | |
XM_011544268.1 | XP_011542570.1 | 13 - 14 | 3010 | 84 | |
XM_011544269.2 | XP_011542571.1 | 11 - 12 | 2668 | 84 | |
XM_017002220.1 | XP_016857709.1 | 15 - 16 | 3290 | 84 | |
GenBank mRNA | AJ318888.2 | - | 14 - 15 | 3094 | 84 |
AK294432.1 | - | 14 - 15 | 3001 | 84 | |
AK294654.1 | - | 4 - 5 | 1085 | 84 | |
AK294964.1 | - | 7 - 8 | 2169 | 84 | |
AK295378.1 | - | 8 - 9 | 1873 | 84 | |
AK316016.1 | - | 7 - 8 | 2071 | 84 | |
AK316103.1 | - | 7 - 8 | 2049 | 84 | |
AL357233.1 | - | 2 - 3 | 323 | 84 | |
BC040247.1 | - | 12 - 13 | 2625 | 84 | |
BC058918.1 | - | 14 - 15 | 3107 | 84 | |
U09582.1 | - | 3 - 4 | 1044 | 84 | |
U58334.1 | - | 15 - 16 | 3226 | 84 |