Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015140.3 | NP_055955.1 | 9 - 10 | 1402 | 63 |
GenBank mRNA | BC001070.2 | - | 9 - 10 | 1366 | 63 |
CT841516.1 | - | 9 - 10 | 1343 | 63 | |
D63487.1 | - | 9 - 10 | 1324 | 63 |