Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004654.3 | NP_004645.2 | 29 - 30 | 5178 | 94 |
XM_017030078.1 | XP_016885567.1 | 28 - 29 | 4333 | 94 | |
GenBank mRNA | AF000986.2 | - | 29 - 30 | 5178 | 94 |
Y13618.1 | - | 27 - 28 | 4290 | 94 | |
Y13619.1 | - | 27 - 28 | 4290 | 94 |