Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004654.3 | NP_004645.2 | 3 - 4 | 1037 | 73 |
XM_017030078.1 | XP_016885567.1 | 2 - 3 | 177 | 73 | |
GenBank mRNA | AF000986.2 | - | 3 - 4 | 1037 | 73 |
Y13618.1 | - | 1 - 2 | 149 | 73 | |
Y13619.1 | - | 1 - 2 | 149 | 73 |