Assay Design



Gene Transcripts

Gene Symbol
RFFL
Entrez Gene ID
117584
Gene Name
ring finger and FYVE-like domain containing E3 ubiquitin protein ligase
Gene Aliases
CARP-2, CARP2, FRING, RIFIFYLIN, RNF189, RNF34L
Chromosome Location
Chr.17: 35009112 - 35089329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017368.1 NP_001017368.1 1 - 2 160 63
GenBank mRNA CR933651.1 - 1 - 2 160 63

View Full Product Details