Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030006.1 | NP_001025177.1 | 11 - 12 | 1632 | 73 |
NM_001282.2 | NP_001273.1 | 11 - 12 | 1632 | 73 | |
XM_005257937.3 | XP_005257994.1 | 11 - 12 | 1697 | 73 | |
XM_005257938.2 | XP_005257995.1 | 11 - 12 | 1598 | 73 | |
XM_005257941.2 | XP_005257998.1 | 11 - 12 | 1567 | 73 | |
XM_011524448.1 | XP_011522750.1 | 11 - 12 | 1996 | 73 | |
XM_011524449.2 | XP_011522751.1 | 11 - 12 | 1697 | 73 | |
XM_011524450.1 | XP_011522752.1 | 11 - 12 | 1598 | 73 | |
XM_011524451.1 | XP_011522753.1 | 11 - 12 | 1567 | 73 | |
XM_011524452.1 | XP_011522754.1 | 12 - 13 | 1765 | 73 | |
XM_011524453.1 | XP_011522755.1 | 12 - 13 | 1683 | 73 | |
XM_011524454.1 | XP_011522756.1 | 12 - 13 | 1699 | 73 | |
XM_011524455.2 | XP_011522757.1 | 12 - 13 | 1683 | 73 | |
XM_017024284.1 | XP_016879773.1 | 11 - 12 | 1697 | 73 | |
XM_017024285.1 | XP_016879774.1 | 12 - 13 | 1765 | 73 | |
XM_017024286.1 | XP_016879775.1 | 12 - 13 | 1730 | 73 | |
XM_017024287.1 | XP_016879776.1 | 11 - 12 | 1697 | 73 | |
GenBank mRNA | AK289528.1 | - | 11 - 12 | 1573 | 73 |
AK292531.1 | - | 11 - 12 | 1510 | 73 | |
AK301522.1 | - | 11 - 12 | 1467 | 73 | |
AK304126.1 | - | 7 - 8 | 987 | 73 | |
AY341427.1 | - | 12 - 13 | 1667 | 73 | |
BC006201.2 | - | 11 - 12 | 1539 | 73 | |
BC012150.1 | - | 2 - 3 | 260 | 73 | |
CR749392.1 | - | 11 - 12 | 1615 | 73 | |
M34175.1 | - | 11 - 12 | 1620 | 73 |