Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282.2 | NP_001273.1 | 14 - 15 | 2175 | 104 |
XM_011524451.1 | XP_011522753.1 | 14 - 15 | 2110 | 104 | |
XM_011524455.2 | XP_011522757.1 | 15 - 16 | 2226 | 104 | |
XM_017024284.1 | XP_016879773.1 | 14 - 15 | 2240 | 104 | |
XM_017024286.1 | XP_016879775.1 | 15 - 16 | 2273 | 104 | |
GenBank mRNA | AY341427.1 | - | 15 - 16 | 2210 | 104 |
M34175.1 | - | 14 - 15 | 2163 | 104 |