Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019114.4 | NP_061987.3 | 17 - 18 | 2271 | 78 |
XM_011518790.1 | XP_011517092.1 | 17 - 18 | 1768 | 78 | |
XM_011518791.2 | XP_011517093.1 | 17 - 18 | 1753 | 78 | |
XM_011518792.2 | XP_011517094.1 | 17 - 18 | 1753 | 78 | |
XM_011518794.2 | XP_011517096.1 | 17 - 18 | 1753 | 78 | |
XM_017014813.1 | XP_016870302.1 | 17 - 18 | 1753 | 78 | |
XM_017014814.1 | XP_016870303.1 | 17 - 18 | 1753 | 78 | |
XM_017014815.1 | XP_016870304.1 | 17 - 18 | 1753 | 78 | |
GenBank mRNA | AB209187.1 | - | 9 - 10 | 863 | 78 |
AF153416.1 | - | 17 - 18 | 2043 | 78 |