Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019114.4 | NP_061987.3 | 18 - 19 | 2395 | 99 |
XM_011518790.1 | XP_011517092.1 | 18 - 19 | 1892 | 99 | |
XM_011518791.2 | XP_011517093.1 | 18 - 19 | 1877 | 99 | |
XM_011518792.2 | XP_011517094.1 | 18 - 19 | 1877 | 99 | |
XM_011518794.2 | XP_011517096.1 | 18 - 19 | 1877 | 99 | |
XM_017014813.1 | XP_016870302.1 | 18 - 19 | 1877 | 99 | |
XM_017014814.1 | XP_016870303.1 | 18 - 19 | 1877 | 99 | |
XM_017014815.1 | XP_016870304.1 | 18 - 19 | 1877 | 99 | |
GenBank mRNA | AB209187.1 | - | 10 - 11 | 987 | 99 |
AF153416.1 | - | 18 - 19 | 2167 | 99 |