Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022842.4 | NP_073753.3 | 2 - 3 | 430 | 85 |
NM_178181.2 | NP_835488.1 | 2 - 3 | 430 | 85 | |
XM_011534024.1 | XP_011532326.1 | 2 - 3 | 388 | 85 | |
XM_017007070.1 | XP_016862559.1 | 2 - 3 | 416 | 85 | |
GenBank mRNA | AF468010.1 | - | 2 - 3 | 405 | 85 |
AK026622.1 | - | 2 - 3 | 405 | 85 | |
AK314441.1 | - | 2 - 3 | 394 | 85 | |
AY026461.1 | - | 2 - 3 | 377 | 85 | |
AY167484.1 | - | 2 - 3 | 387 | 85 | |
AY358779.1 | - | 2 - 3 | 388 | 85 | |
AY375452.1 | - | 2 - 3 | 376 | 85 | |
BC021099.1 | - | 2 - 3 | 430 | 85 | |
BC069254.1 | - | 2 - 3 | 383 | 85 | |
KU215672.1 | - | 1 - 2 | 208 | 85 | |
KU257676.1 | - | 1 - 2 | 205 | 85 |