Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022842.4 | NP_073753.3 | 3 - 4 | 796 | 65 |
NM_178181.2 | NP_835488.1 | 3 - 4 | 796 | 65 | |
XM_011534024.1 | XP_011532326.1 | 3 - 4 | 754 | 65 | |
XM_017007070.1 | XP_016862559.1 | 3 - 4 | 782 | 65 | |
GenBank mRNA | AF468010.1 | - | 3 - 4 | 771 | 65 |
AK023834.1 | - | 1 - 2 | 374 | 65 | |
AK026622.1 | - | 3 - 4 | 771 | 65 | |
AK314441.1 | - | 3 - 4 | 760 | 65 | |
AY026461.1 | - | 3 - 4 | 743 | 65 | |
AY167484.1 | - | 3 - 4 | 753 | 65 | |
AY358779.1 | - | 3 - 4 | 754 | 65 | |
AY375452.1 | - | 3 - 4 | 742 | 65 | |
BC021099.1 | - | 3 - 4 | 796 | 65 | |
BC069254.1 | - | 3 - 4 | 749 | 65 | |
KU215672.1 | - | 2 - 3 | 574 | 65 | |
KU257676.1 | - | 2 - 3 | 571 | 65 |