Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015255.2 | NP_056070.1 | 25 - 26 | 3064 | 88 |
XM_005248965.4 | XP_005249022.1 | 25 - 26 | 3181 | 88 | |
XM_005248966.3 | XP_005249023.1 | 24 - 25 | 2464 | 88 | |
XM_011514438.2 | XP_011512740.2 | 25 - 26 | 3264 | 88 | |
XM_011514439.1 | XP_011512741.1 | 16 - 17 | 1631 | 88 | |
XM_011514440.1 | XP_011512742.1 | 16 - 17 | 1631 | 88 | |
XM_011514441.2 | XP_011512743.1 | 13 - 14 | 1244 | 88 | |
XM_017010594.1 | XP_016866083.1 | 25 - 26 | 3265 | 88 | |
XM_017010595.1 | XP_016866084.1 | 25 - 26 | 3265 | 88 | |
XM_017010596.1 | XP_016866085.1 | 25 - 26 | 3099 | 88 | |
XM_017010597.1 | XP_016866086.1 | 25 - 26 | 3266 | 88 | |
GenBank mRNA | AB002347.1 | - | 13 - 14 | 1326 | 88 |
AK026998.1 | - | 4 - 5 | 327 | 88 | |
AK125795.1 | - | 17 - 18 | 1730 | 88 | |
AK304070.1 | - | 17 - 18 | 1717 | 88 | |
AY061884.1 | - | 25 - 26 | 2766 | 88 | |
BX647467.1 | - | 8 - 9 | 737 | 88 |