Assay Design



Gene Transcripts

Gene Symbol
LIPC
Entrez Gene ID
3990
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
Chromosome Location
Chr.15: 58410754 - 58568952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000236.2 NP_000227.2 3 - 4 507 62
XM_005254372.1 XP_005254429.1 4 - 5 555 62
XM_005254374.4 XP_005254431.2 3 - 4 1849 62
XM_006720502.3 XP_006720565.1 3 - 4 452 62
XM_017022176.1 XP_016877665.1 3 - 4 1864 62
GenBank mRNA AK290062.1 - 5 - 6 1058 62
AK292631.1 - 5 - 6 1061 62
AK309448.1 - 2 - 3 307 62
AK315306.1 - 3 - 4 686 62
BC132825.1 - 3 - 4 485 62
BC136495.1 - 3 - 4 485 62
BC146659.1 - 4 - 5 730 62
D83548.1 - 3 - 4 491 62
J03540.1 - 3 - 4 454 62
J03895.1 - 3 - 4 457 62
X07228.1 - 3 - 4 507 62

View Full Product Details