Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165977.2 | NP_001159449.1 | 5 - 6 | 818 | 58 |
NM_001165978.2 | NP_001159450.1 | 5 - 6 | 818 | 58 | |
NM_001321070.1 | NP_001307999.1 | 5 - 6 | 879 | 58 | |
NM_144707.3 | NP_653308.2 | 5 - 6 | 818 | 58 | |
XM_011510672.2 | XP_011508974.1 | 5 - 6 | 802 | 58 | |
GenBank mRNA | AF245303.1 | - | 5 - 6 | 813 | 58 |
AF245304.1 | - | 5 - 6 | 813 | 58 | |
AK091175.1 | - | 5 - 6 | 788 | 58 | |
AK290366.1 | - | 5 - 6 | 773 | 58 | |
AK313267.1 | - | 5 - 6 | 774 | 58 | |
AY358377.1 | - | 5 - 6 | 769 | 58 | |
BC114525.1 | - | 5 - 6 | 793 | 58 |