Assay Design



Gene Transcripts

Gene Symbol
PKD1L1
Entrez Gene ID
168507
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
Gene Aliases
PRO19563
Chromosome Location
Chr.7: 47774603 - 47958622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138295.3 NP_612152.1 35 - 36 5587 74
XM_017011798.1 XP_016867287.1 36 - 37 11992 74
GenBank mRNA AB061683.1 - 35 - 36 5553 74

View Full Product Details