Assay Design
Gene Transcripts
- Gene Symbol
- CLCN6
- Entrez Gene ID
-
1185
- Gene Name
- chloride voltage-gated channel 6
- Gene Aliases
-
CLC-6
- Chromosome Location
-
Chr.1: 11806096 - 11843144 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X99475.1 |
- |
9 - 10 |
707 |
88 |
View Full Product Details