Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013369.3 | NP_037501.2 | 2 - 3 | 592 | 78 |
NM_175867.2 | NP_787063.1 | 2 - 3 | 592 | 78 | |
GenBank mRNA | AF194032.1 | - | 2 - 3 | 592 | 78 |
BC002560.2 | - | 2 - 3 | 266 | 78 |