Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004533.3 | NP_004524.3 | 22 - 23 | 2650 | 72 |
GenBank mRNA | BC130536.1 | - | 22 - 23 | 2617 | 72 |
BC136389.1 | - | 22 - 23 | 2617 | 72 | |
X73113.1 | - | 22 - 23 | 2621 | 72 |