Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000823.3 | NP_000814.2 | 8 - 9 | 863 | 86 |
XM_011515263.1 | XP_011513565.1 | 5 - 6 | 881 | 86 | |
GenBank mRNA | AB058895.1 | - | 3 - 4 | 391 | 86 |
AF282259.1 | - | 5 - 6 | 881 | 86 | |
AF282260.1 | - | 4 - 5 | 727 | 86 | |
AF282261.1 | - | 5 - 6 | 551 | 86 | |
AF282262.1 | - | 2 - 3 | 496 | 86 | |
AV750179.1 | - | 3 - 4 | 86 | ||
AY557192.1 | - | 8 - 9 | 815 | 86 | |
L01406.1 | - | 8 - 9 | 866 | 86 | |
L09237.1 | - | 8 - 9 | 863 | 86 |