Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000823.3 | NP_000814.2 | 6 - 7 | 640 | 92 |
XM_011515263.1 | XP_011513565.1 | 3 - 4 | 658 | 92 | |
GenBank mRNA | AF282259.1 | - | 3 - 4 | 658 | 92 |
AV750179.1 | - | 1 - 2 | 62 | 92 | |
AY557192.1 | - | 6 - 7 | 592 | 92 | |
L09237.1 | - | 6 - 7 | 640 | 92 |