Assay Design



Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
Chromosome Location
Chr.2: 219171897 - 219178174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321390.1 NP_001308319.1 1 - 2 60 63

View Full Product Details