Assay Design
Gene Transcripts
- Gene Symbol
- MBD6
- Entrez Gene ID
-
114785
- Gene Name
- methyl-CpG binding domain protein 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 57520710 - 57530148 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL834230.1 |
- |
8 - 9 |
1674 |
68 |
View Full Product Details